EMAIL THIS PAGE TO A FRIEND
Stem Cells and Development
Embryonic Stem Cell-Related Protein L1TD1 Is Required for Cell Viability, Neurosphere Formation, and Chemoresistance in Medulloblastoma.
PMID 26159230
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU019341 MISSION® esiRNA, esiRNA targeting human L1TD1 (esiRNA1)