EMAIL THIS PAGE TO A FRIEND
Nature communications
PGD2 deficiency exacerbates food antigen-induced mast cell hyperplasia.
PMID 26159556
Abstract
Prostaglandin D2 (PGD2) is a major prostanoid secreted mainly by mast cells. Although PGD2 has been identified as a modulator of allergic inflammation, its precise role remains unclear. Here we investigate the role of PGD2 in food allergy. Oral administration of ovalbumin induces allergic responses in sensitized wild-type (WT) mice. Systemic gene deficiency of haematopoietic PGD synthase (H-PGDS(-/-)) exacerbates all of the manifestations accompanying severe mast cell hyperplasia in the intestine. Morphological studies show that c-kit/FcɛRI-positive WT mast cells strongly express H-PGDS. Transplantation of H-PGDS(-/-) mast cells also aggravates ovalbumin-induced mast cell hyperplasia and allergic symptoms in mast cell null mice. H-PGDS deficiency accelerates the production of SDF-1α and the activity of MMP-9 in the antigen-stimulated intestine. SDF-1α receptor blockade or MMP-9 inhibition relieves the exacerbated mast cell hyperplasia and manifestations observed in H-PGDS(-/-). Thus, PGD2 deficiency results in food antigen-induced mast cell hyperplasia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3766
Thymine 1-β-D-arabinofuranoside, ≥99%
C10H14N2O6