EMAIL THIS PAGE TO A FRIEND
Journal of applied physiology (Bethesda, Md. : 1985)
Recording of intracranial pressure in conscious rats via telemetry.
PMID 26159754
Abstract
Although cerebral perfusion pressure (CPP) is known to be fundamental in the control of normal brain function, there have been no previous long-term measurements in animal models. The aim of this study was to explore the stability and viability of long-term recordings of intracranial pressure (ICP) in freely moving rats via a telemetry device. We also developed a repeatable surgical approach with a solid-state pressure sensor at the tip of the catheter placed under the dura and in combination with arterial pressure (AP) measurement to enable the calculation of CPP. Telemeters with dual pressure catheters were implanted in Wistar rats to measure ICP and AP. We found that the signals were stable throughout the 28-day recording period with an average ICP value of 6 ± 0.8 mmHg. Significant light-dark differences were found in AP (3.1 ± 2.7 mmHg, P = 0.02) and HR (58 ± 12 beats/min, P = 0.003), but not ICP (0.3 ± 0.2 mmHg, P >0.05) or CPP (2.6 ± 2.8 mmHg, P > 0.05). Use of kaolin to induce hydrocephalus in several rats demonstrates the ability to measure changes in ICP throughout disease progression, validating this new solution for chronic measurement of ICP, CPP, and AP in conscious rats.