EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Pyrene is a Novel Constitutive Androstane Receptor (CAR) Activator and Causes Hepatotoxicity by CAR.
PMID 26160115
Abstract
Polycyclic aromatic hydrocarbons (PAHs) are a class of ubiquitous persistent environmental pollutants which are primarily formed from the incomplete combustion of organic materials. Many potential sources of human exposure to PAHs exist, including daily exposures from the ambient environment or occupational settings. PAHs have been found to cause harmful effects on human health. Here, we evaluated the adverse effects of pyrene, a common PAH, on the liver. The present study demonstrates that pyrene is able to activate mouse constitutive androstane receptor (CAR). CAR protein, as measured by Western blot analysis, was observed to translocate into the nucleus from the cytoplasm in mouse liver after exposure to pyrene. Utilizing CAR null mice, we identified that CAR mediates pyrene-induced hepatotoxicity. Increased relative liver weight, hepatocellular hypertrophy, and elevated serum alanine aminotransferase levels were found in wild-type but not CAR null mice after orally administered pyrene. We further show that pyrene induced the expression of mouse liver metabolism enzymes including CYP2B10, CYP3A11, GSTm1, GSTm3, and SULT1A1, and caused hepatic glutathione depletion in wild-type but not CAR null mice. Moreover, by luciferase reporter assay and quantitative real-time PCR analysis, pyrene was found to be a potential inducer of CYP2B6 expression via activation of human CAR in HepG2 cells and human primary hepatocytes. Our observations suggest that pyrene is a novel CAR activator and that CAR is essential for mediating pyrene-induced liver injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1756
Dexamethasone, ≥98% (HPLC), powder
C22H29FO5
D4902
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
C22H29FO5
D8893
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
C22H29FO5
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5
G6529
L-Glutathione reduced, BioXtra, ≥98.0%
C10H17N3O6S
G6013
L-Glutathione reduced, cell culture tested, BioReagent, ≥98.0%, powder
C10H17N3O6S
P3532
Phenol Red, powder, BioReagent, suitable for cell culture
C19H14O5S
114529
Phenol Red, ACS reagent
C19H14O5S
484431
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
C6H12O3
537543
Propylene glycol monomethyl ether acetate, 99%
C6H12O3
82648
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
C16H10
571245
Pyrene, sublimed grade, 99%
C16H10
185515
Pyrene, 98%
C16H10
40082
Pyrene, certified reference material, 1000 μg/mL in methanol
C16H10
48570
Pyrene, analytical standard
C16H10
792500
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
C3H3NaO3
80443
Sodium pyruvate, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H3NaO3
P2256
Sodium pyruvate, ReagentPlus®, ≥99%
C3H3NaO3
P8574
Sodium pyruvate, BioXtra, ≥99%
C3H3NaO3
P5280
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
C3H3NaO3
P4562
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo
C3H3NaO3
P3662
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
C3H3NaO3
T1443
TCPOBOP, ≥98% (HPLC), solid
C16H8N2O2Cl4