EMAIL THIS PAGE TO A FRIEND
Circulation
Incidence and Clinical Significance of Poststent Optical Coherence Tomography Findings: One-Year Follow-Up Study From a Multicenter Registry.
PMID 26162917
Abstract
Optical coherence tomography (OCT) was recently introduced to optimize percutaneous coronary intervention. However, the exact incidence and significance of poststent OCT findings are unknown. A total of 900 lesions treated with 1001 stents in 786 patients who had postprocedure OCT imaging were analyzed to evaluate the incidence of poststent OCT findings and to identify the OCT predictors for device-oriented clinical end points, including cardiac death, target vessel-related myocardial infarction, target lesion revascularization, and stent thrombosis. Patients were followed up to 1 year. Stent edge dissection was detected in 28.7% of lesions, and incomplete stent apposition was detected in 39.1% of lesions. The incidences of smooth protrusion, disrupted fibrous tissue protrusion, and irregular protrusion were 92.9%, 61.0%, and 53.8%, respectively. Small minimal stent area, defined as a lesion with minimal stent area <5.0 mm(2) in a drug-eluting stent or <5.6 mm(2) in a bare metal stent, was observed in 40.4% of lesions. One-year device-oriented clinical end points occurred in 33 patients (4.5%). Following adjustment, irregular protrusion and small minimal stent area were independent OCT predictors of 1-year device-oriented clinical end points (P=0.003 and P=0.012, respectively). Abnormal poststent OCT findings were frequent. Irregular protrusion and small minimal stent area were independent predictors of 1-year device-oriented clinical end points, which were primarily driven by target lesion revascularization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774278
Diethyl sulfite, 98%
C4H10O3S