EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Persistent Antigen and Prolonged AKT-mTORC1 Activation Underlie Memory CD8 T Cell Impairment in the Absence of CD4 T Cells.
PMID 26163589
Abstract
Recall responses by memory CD8 T cells are impaired in the absence of CD4 T cells. Although several mechanisms have been proposed, the molecular basis is still largely unknown. Using a local influenza virus infection in the respiratory tract and the lung of CD4(-/-) mice, we show that memory CD8 T cell impairment is limited to the lungs and the lung-draining lymph nodes, where viral Ags are unusually persistent and abundant in these mice. Persistent Ag exposure results in prolonged activation of the AKT-mTORC1 pathway in Ag-specific CD8 T cells, favoring their development into effector memory T cells at the expense of central memory T cells, and inhibition of mTORC1 by rapamycin largely corrects the impairment by promoting central memory T cell development. The findings suggest that the prolonged AKT-mTORC1 activation driven by persistent Ag is a critical mechanism underlying the impaired memory CD8 T cell development and responses in the absence of CD4 T cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0395
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
C51H79NO13