EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Enzyme and Mediator-coadsorbed Carbon Felt Electrode for Electrochemical Detection of Glucose Covered with Polymer Layers Based on Layer-by-Layer Technique.
PMID 26165293
Abstract
Glucose dehydrogenase (GlDH) and ferrocene were coadsorbed on a carbon felt (CF) sheet (5 × 10 mm, 2 mm thickness), which was used to construct an electrode for the electrochemical detection of glucose. A potential of +0.3 V vs. Ag/AgCl was applied on the base CF, and the current was measured. After the addition of glucose, the current increased and reached a steady state within 50 s. The current response was proportional to the glucose concentration up to 20 μM, with a lower detection limit of 1 μM. The surface of the CF electrode was covered by layers of polystyrene sulfonate and poly-L-lysine using layer-by-layer technique. Again the current response was proportional to glucose concentration up to 20 μM, with a lower detection limit of 2 μM. The oxidation current owing to electrochemical interferents such as L-ascorbate and acetaminophen was 1/8 times of the current observed on the unprotected electrode. In addition, the protection imparted stability to the electrode. Our work demonstrates that a GlDH/ferrocene CF electrode, protected with polystyrene sulfonate and poly-L-lysine, could be used for the electrochemical detection of glucose.