EMAIL THIS PAGE TO A FRIEND
PMID 26167517
Abstract
The real time visualisation of fluorescently tagged proteins in live cells using ever more sophisticated microscopes has greatly increased our understanding of the dynamics of key proteins during fundamental physiological processes such as cell locomotion, chemotaxis, cell division and membrane trafficking. In addition the fractionation of cells and isolation of organelles or known compartments can often verify any subcellular localisation and the use of tagged proteins as bait for the immunoprecipitation of material from cell fractions can identify specific binding partners and multiprotein complexes thereby helping assign a function to the tagged protein. We have successfully applied these techniques to the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PWOPOL-RO
Pwo DNA Polymerase