EMAIL THIS PAGE TO A FRIEND
Antimicrobial Agents and Chemotherapy
Cellular pharmacokinetics and intracellular activity of the novel peptide deformylase inhibitor GSK1322322 against Staphylococcus aureus laboratory and clinical strains with various resistance phenotypes: studies with human THP-1 monocytes and J774 murine macrophages.
PMID 26169402