EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
Study of polyethyleneimine- and amidoxime-functionalized hybrid biomass of Spirulina (Arthrospira) platensis for adsorption of uranium (VI) ion.
PMID 26169821