EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
Hypogonadism Associated with Cyp19a1 (Aromatase) Posttranscriptional Upregulation in Celf1 Knockout Mice.
PMID 26169831
Abstract
CELF1 is a multifunctional RNA-binding protein that controls several aspects of RNA fate. The targeted disruption of the Celf1 gene in mice causes male infertility due to impaired spermiogenesis, the postmeiotic differentiation of male gametes. Here, we investigated the molecular reasons that underlie this testicular phenotype. By measuring sex hormone levels, we detected low concentrations of testosterone in Celf1-null mice. We investigated the effect of Celf1 disruption on the expression levels of steroidogenic enzyme genes, and we observed that Cyp19a1 was upregulated. Cyp19a1 encodes aromatase, which transforms testosterone into estradiol. Administration of testosterone or the aromatase inhibitor letrozole partly rescued the spermiogenesis defects, indicating that a lack of testosterone associated with excessive aromatase contributes to the testicular phenotype. In vivo and in vitro interaction assays demonstrated that CELF1 binds to Cyp19a1 mRNA, and reporter assays supported the conclusion that CELF1 directly represses Cyp19a1 translation. We conclude that CELF1 downregulates Cyp19a1 (Aromatase) posttranscriptionally to achieve high concentrations of testosterone compatible with spermiogenesis completion. We discuss the implications of these findings with respect to reproductive defects in men, including patients suffering from isolated hypogonadotropic hypogonadism and myotonic dystrophy type I.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6545
Letrozole, ≥98% (HPLC)
C17H11N5
P8825
Monoclonal Anti-Proliferating Cell Nuclear Antigen antibody produced in mouse, clone PC 10, ascites fluid
746436
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
ClK
793590
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
ClK
P9333
Potassium chloride, BioXtra, ≥99.0%
ClK
P9541
Potassium chloride, for molecular biology, ≥99.0%
ClK
P5405
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
ClK
P1597
Potassium chloride, meets EP, USP testing specifications
ClK
449989
Potassium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClK
451029
Potassium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClK
409316
Potassium chloride, 99.999% trace metals basis
ClK
204099
Potassium chloride, ≥99.99% trace metals basis
ClK
60128
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClK
P9327
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
ClK
60142
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
ClK
44675
Potassium chloride solution, 1 M
ClK
T1500
Testosterone, ≥98%
C19H28O2
T1875
Testosterone propionate, solid
C22H32O3
T-037
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C19H28O2