EMAIL THIS PAGE TO A FRIEND
The Journal of Steroid Biochemistry and Molecular Biology
Comparative analysis of intraluteal steroidogenic enzymes emphasises the functionality of fresh and persistent corpora lutea during pro--and metoestrus in the lynx.
PMID 26170243
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7567
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
C12H27P