EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
The effects of okra (Abelmoschus esculentus Linn.) on the cellular events associated with Alzheimer's disease in a stably expressed HFE neuroblastoma SH-SY5Y cell line.
PMID 26170247
Abstract
It has been reported that persons carrying the H63D variant in their hemochromatosis (HFE) gene are at increased risk of Alzheimer's disease (AD). We investigated the possibility that okra (Abelmoschus esculentus) and quercetin could mitigate this risk factor by examining its effect on AD-associated cellular events in HFE stably expressing SH-SY5Y cells. Treatment of H63D HFE cells either with okra or quercetin significantly decreased reactive oxygen species (ROS), hydrogen peroxide (H2O2), and protein oxidation compared to untreated cells. The levels of tau phosphorylation at serine-199, serine-202, and serine-396 sites were also significantly decreased when cells were treated with okra. Exposure of the H63D and wild type (WT) cells to iron increased tau phosphorylation, but this response was decreased significantly when cells were treated with okra. The mechanism responsible for these changes appears to be related to decreased glycogen synthase kinase (GSK)-3β activity, an upstream signaling kinase of tau phosphorylation. We also established that okra treatment dramatically decreases intracellular iron levels in H63D cells compared to untreated cells. Our results provide important in vitro data on the effects of okra on various AD-associated cellular processes in H63D variant HFE cells. These results suggest okra may be beneficial in people expressing the H63D variant to reduce the risk of AD and other neurodegenerative diseases related to oxidative stress. Further in vivo studies would help confirm this.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17793
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
C21H20O12