EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Role of Bcl-2/adenovirus E1B 19 kDa-interacting protein 3 in myocardial cells in diabetes.
PMID 26170914
Abstract
Cardiovascular complications are the major causes of morbidity and mortality associated with Type 2 Diabetes. Among the macrovascular complications, diabetic cardiomyopathy (DCM) is generally considered to be inadequately recognized and managed. Bcl-2/adenovirus E1B 19 kDa-interacting protein 3 (BNIP3), is known to play a key role in the initiation of the mitochondrial pathway of apoptosis induced by hypoxia and acidosis in the heart. It is unknown whether BNIP3 is also important for cardiac cell survival or adaption in response to hyperglycemia. Based on the previous finding that BNIP3 was significantly induced in the diabetic rat heart, BNIP3 was transfected in primary rat cardiomyocytes and the H9c2 cell line in the present study. Overexpressed BNIP3 decreased the mitochondrial membrane potential and induced cell apoptosis. When BNIP3 was knocked down, the effect on cell apoptosis was reversed. Transcriptome analysis showed that the genes regulating mitochondrial metabolism, such as carnitine palmitoyltransferase 1b, cytochrome