EMAIL THIS PAGE TO A FRIEND
Oncology letters
Proflavin suppresses the growth of human osteosarcoma MG63 cells through apoptosis and autophagy.
PMID 26171052
Abstract
Proflavin is one of the novel acridine derivatives that possess various pharmacological effects. Although numerous studies have been performed to investigate proflavin, its effects have not been investigated on the human osteosarcoma MG63 cell line. The core aim of the present study was to test the effects of proflavin on the viability of MG63 cells and the induction of apoptosis and autophagy in MG63 cells. The induction of apoptosis was examined by measuring the changes in the expression of the B-cell lymphoma 2 (Bcl-2) and Bcl-2-associated X protein mRNA and proteins. Apoptotic cell death was identified by the proteolytic cleavage of poly (adenosine diphosphate-ribose) polymerase and caspase-3 and caspase-9. In addition, the autophagic effects of proflavin were examined by the quantitation of the mRNA expression of autophagy protein 5 and Beclin 1, in addition to the identification of the accumulation of microtubule-associated protein 1 light chain 3-II. The present results revealed that proflavin inhibited the proliferation of MG63 cells in a dose-dependent manner. Proflavin-induced cell death was attributed to apoptosis and autophagy. Overall, the present results indicated that the antiseptic agent proflavin exerts anticancer potential through the synergistic activity of apoptosis and autophagy.