EMAIL THIS PAGE TO A FRIEND
Polish journal of veterinary sciences
Caudal mesenteric ganglion in the sheep - macroanatomical and immunohistochemical study.
PMID 26172189
Abstract
The caudal mesenteric ganglion (CaMG) is a prevetrebral ganglion which provides innervation to a number of organs in the abdominal and pelvic cavity. The morphology of CaMG and the chemical coding of neurones in this ganglion have been described in humans and many animal species, but data on this topic in the sheep are entirely lacking. This prompted us to undertake a study to determine the localization and morphology of sheep CaMG as well as immunohistochemical properties of its neurons. The study was carried out on 8 adult sheep, weighing from 40 to 60 kg each. The sheep were deeply anaesthetised and transcardially perfused with 4% paraformaldehyde. CaMG-s were exposed and their location was determined. Macroanatomical observations have revealed that the ovine CaMG is located at the level of last two lumbar (L5 or L6) and the first sacral (S1) vertebrae. The ganglion represents an unpaired structure composed of several, sequentially arranged aggregates of neurons. Immunohistochemical investigations revealed that nearly all (99.5%) the neurons were DβH-IR and were richly supplied by VACHT-IR nerve terminals forming "basket-like" structures around the perikarya. VACHT-IR neurones were not determined. Many neurons (55%) contained immunoreactivity to NPY, some of them (10%) stained for Met-ENK and solitary nerve cells were GAL-positive. CGRP-IR nerve fibres were numerous and a large number of them simultaneously expressed immunoreactivity to SP. Single, weakly stained neurones were SP-IR and only very few nerve cells weakly stained for VIP.