EMAIL THIS PAGE TO A FRIEND
Journal of neuropathology and experimental neurology
Evidence for Epigenetic Regulation of Gene Expression and Function in Chronic Experimental Diabetic Neuropathy.
PMID 26172287
Abstract
Diabetic polyneuropathy (DPN) is a common but irreversible neurodegenerative complication of diabetes mellitus. Here we show that features of sensory neuron damage in mice with chronic DPN may have altered epigenetic micro RNA (miRNA) transcriptional control. We profiled sensory neuron messenger RNA and miRNA profiles in mice with type I diabetes mellitus and findings of DPN. Diabetic sensory dorsal root ganglia neurons showed a pattern of altered messenger RNA profiles associated with upregulated cytoplasmic sites of miRNA-mediated messenger RNA processing (GW/P bodies). Dorsal root ganglia miRNA microarray identified significant changes in expression among mice with diabetes, the most prominent of which were a 39% downregulation of mmu-let-7i and a 255% increase in mmu-miR-341; both were identified in sensory neurons. To counteract these alterations, we replenished let-7i miRNA by intranasal administration; in a separate experiment, we added an anti-miR that antagonized elevated mmu-341 after 5 months of diabetes. Both approaches independently improved electrophysiologic, structural, and behavioral abnormalities without altering hyperglycemia; control sequences did not have these effects. Dissociated adult sensory neurons exposed to an exogenous mmu-let-7i mimic displayed enhanced growth and branching, indicating a trophic action. These findings identify roles for epigenetic miRNA alterations and enhanced GW/P expression in diabetic dorsal root ganglia that contribute to the complex DPN phenotype.