EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Fluorescent Phosphorus Dendrimer as a Spectral Nanosensor for Macrophage Polarization and Fate Tracking in Spinal Cord Injury.
PMID 26175127
Abstract
Dendrimers and dendriplexes, highly branched synthetic macromolecules, have gained popularity as new tools for a variety of nanomedicine strategies due to their unique structure and properties. We show that fluorescent phosphorus dendrimers are well retained by bone marrow-derived macrophages and exhibit robust spectral shift in its emission in response to polarization conditions. Fluorescence properties of this marker can also assist in identifying macrophage presence and phenotype status at different time points after spinal cord injury. Potential use of a single dendrimer compound as a drug/siRNA carrier and phenotype-specific cell tracer offers new avenues for enhanced cell therapies combined with monitoring of cell fate and function in spinal cord injury.