EMAIL THIS PAGE TO A FRIEND
Stem Cells and Development
Activation of Type 4 Metabotropic Glutamate Receptor Attenuates Oxidative Stress-Induced Death of Neural Stem Cells with Inhibition of JNK and p38 MAPK Signaling.
PMID 26176363
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1640
VU0155041, ≥98% (HPLC)
C14H15Cl2NO3