EMAIL THIS PAGE TO A FRIEND
Journal of food science
Microbial Safety and Shelf Life of UV-C Treated Freshly Squeezed White Grape Juice.
PMID 26177986
Abstract
The effects of UV-C irradiation on the inactivation of Escherichia coli K-12 (ATCC 25253), a surrogate of E. coli O157:H7, and on the shelf life of freshly squeezed turbid white grape juice (FSWGJ) were investigated. FSWGJ samples were processed at 0.90 mL/s for 32 min by circulating 8 times in an annular flow UV system. The UV exposure time was 244 s per cycle. The population of E. coli K-12 was reduced by 5.34 log cycles after exposure to a total UV dosage of 9.92 J/cm(2) (1.24 J/cm(2) per cycle) at 0.90 mL/s flow rate. The microbial shelf life of UV-C treated FSWGJ was extended up to 14 d at 4 °C. UV exposure was not found to alter pH, total soluble solid, and titratable acidity of juice. There was a significant effect (P < 0.05) on turbidity, absorbance coefficient, color, and ascorbic acid content. Furthermore, all physicochemical properties were altered during refrigerated storage. The microbial shelf life of FSWGJ was doubled after UV-C treatment, whereas the quality of juice was adversely affected similarly observed in the control samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
438464
Potassium iodate, 99.995% trace metals basis
IKO3