EMAIL THIS PAGE TO A FRIEND
Cardiovascular Drugs and Therapy
Cardioprotective Properties of the Platelet P2Y12 Receptor Inhibitor, Cangrelor: Protective in Diabetics and Reliant Upon the Presence of Blood.
PMID 26179955