EMAIL THIS PAGE TO A FRIEND
European journal of mass spectrometry (Chichester, England)
New insights into coenzyme A interaction with mercury ions provided by mass spectrometric and circular dichroism spectroscopic approaches.
PMID 26181283
Abstract
The interaction of coenzyme A (CoA) with mercury ions was investigated using electrospray ionization mass spectrometry and circular dichroism spectroscopy. Our results indicated a 1:1 stoichiometric CoA-Hg complex at physiological pH. Furthermore, the CoA conformation considerably changed in the presence of mercury ions. In addition, a by-product of the reaction, thiocoenzyme A, was identified using mass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
413445
Mercury(II) chloride, ≥98%
Cl2Hg