EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Effect of drying methods on the free and conjugated bufadienolide content in toad venom determined by ultra-performance liquid chromatography-triple quadrupole mass spectrometry coupled with a pattern recognition approach.
PMID 26186722
Abstract
Drying is a useful technique for extending the shelf-life of biological products and enabling long-term storage; however, improper drying can reduce the chemical quality of the products. In this study, we used ultra-performance liquid chromatography-triple quadrupole/mass spectrometry (LC-MS/MS) and multivariate statistical analysis to investigate the effects of four drying methods (V: vacuum-drying at 60°C, F: freeze-drying, H: air-drying at 60°C and R: air-drying at room temperature) on the levels of 36 bufadienolides in toad venom. Vacuum-drying at 60°C produced the highest quality dried toad venom in terms of total bufadienolide content, whereas traditional air-drying at room temperature (RT) to dehydrate the toad venom led to a dramatic loss in free and conjugated bufadienolides, reaching up to 60% and 70%, respectively. Assaying for free bufadienolides ranked the drying methods as V≈F>H>R, whereas assaying for conjugated bufadienolides slightly changed the order to V>F≈H>R. Furthermore, we identified 21 bufadienolides as biomarkers responsible for the decline in the quality of dried toad venom, whose loss varied from 1.5-fold to 100-fold. Of these biomarkers, group I bufadienolides that contain 16-OAc (e.g., cinobufagin and its hydroxyl or arginine ester derivatives) were characteristic components and were reduced to trace levels (loss of more than 10-fold) following traditional air-drying at RT. This might be attributed to the fact that most enzyme-sensitive bufadienolides were biotransformed or degraded at room temperature but were retained using other drying methods.