EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Polyamidoamine (PAMAM) dendrimers modified with short oligopeptides for early endosomal escape and enhanced gene delivery.
PMID 26187169
Abstract
Recently, non-viral vectors have become a popular research topic in the field of gene therapy. In this study, we conjugated short oligopeptides to polyamidoamine-generation 4 (PAMAM G4) to achieve higher transfection efficiency. Previous reports have shown that the PAMAM G4-histidine (H)-arginine (R) dendrimer enhances gene delivery by improving cell penetration and internalization mechanisms. Therefore, we synthesized PAMAM G4-H phenylalanine (F) R, PAMAM G4-FHR and PAMAM G4-FR derivatives to determine the best gene carrier with the lowest toxicity. Physicochemical studies were performed to determine mean diameters and surface charge of PAMAM derivatives/pDNA polyplexes. DNA condensation was confirmed using a gel retardation assay. Cytotoxicity and transfection efficiency were analyzed using human cervical carcinoma (HeLa) and human liver carcinoma (HepG2) cells. Similar levels of transfection were achieved in both cell lines by using gold standard transfection reagent PEI 25 kD. Therefore, our results show that these carriers are promising and may help achieve higher transfection with negligible cytotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
383767
Tri(2-furyl)phosphine, 99%
C12H9O3P