EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Impaired oxidative phosphorylation regulates necroptosis in human lung epithelial cells.
PMID 26187663
Abstract
Cellular metabolism can impact cell life or death outcomes. While metabolic dysfunction has been linked to cell death, the mechanisms by which metabolic dysfunction regulates the cell death mode called necroptosis remain unclear. Our study demonstrates that mitochondrial oxidative phosphorylation (OXPHOS) activates programmed necrotic cell death (necroptosis) in human lung epithelial cells. Inhibition of mitochondrial respiration and ATP synthesis induced the phosphorylation of mixed lineage kinase domain-like protein (MLKL) and necroptotic cell death. Furthermore, we demonstrate that the activation of AMP-activated protein kinase (AMPK), resulting from impaired mitochondrial OXPHOS, regulates necroptotic cell death. These results suggest that impaired mitochondrial OXPHOS contributes to necroptosis in human lung epithelial cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A59654
5-Aminoindole, 97%
C8H8N2