EMAIL THIS PAGE TO A FRIEND
Food microbiology
Cold plasma treatment for the microbiological safety of cabbage, lettuce, and dried figs.
PMID 26187830
Abstract
Microwave-powered cold plasma treatment (CPT) was evaluated as a means to improve the microbiological safety of fresh vegetables and dried fruits. The CPT at 900xa0W, conducted for 10xa0min using nitrogen as a plasma-forming gas, inactivated Salmonella Typhimurium inoculated on cabbage and lettuce by approximately 1.5xa0log CFU/g. The CPT at 400-900xa0W and 667xa0Pa, conducted for 1-10xa0min using a helium-oxygen gas mixture, inactivated Listeria monocytogenes on cabbage by 0.3-2.1xa0log CFU/g in a time-dependent manner (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
705233
1-Decanethiol, 99%
C10H22S