EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Anti-Plasmodium falciparum activity of quinoline-sulfonamide hybrids.
PMID 26190461
Abstract
Fifteen quinoline-sulfonamide hybrids, with a 7-chloroquinoline moiety connected by a linker group to arylsulfonamide moieties with different substituents in the 4-position were synthesized and assayed against Plasmodium falciparum. The compounds displayed high schizonticidal blood activity in vitro, with IC50 values ranging from 0.05 to 1.63 μM, in the anti-HPR2 assay against clone W2-chloroquine-resistant; ten of them showed an IC50 (ranging from 0.05 to 0.40 μM) lower than that of chloroquine and sulfadoxine. Among them, two compounds inhibited Plasmodium berghei parasitemia by 47% and 49% on day 5 after mice inoculation. The most active, in vivo, hybrid 13 is considered to be a new prototype for the development of an antimalarial drug against chloroquine-resistant parasites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7821
Sulfadoxin, ≥95% (TLC)
C12H14N4O4S