EMAIL THIS PAGE TO A FRIEND
Food & function
Promising new applications of Castanea sativa shell: nutritional composition, antioxidant activity, amino acids and vitamin E profile.
PMID 26190821
Abstract
The present study was aimed to assess the macronutrient composition and the amino acid and vitamin E profiles of Castanea sativa shell from different production regions of Portugal (Minho, Trás-os-Montes and Beira-Alta). The nutritional composition was similar for all samples, with a high moisture content and low fat amounts. Arginine and leucine were the predominant essential amino acids (EAA) accounting for 3.55-7.21% and 1.59-2.08%, respectively, for samples of the different production zones. All the shells presented high contents of vitamin E (481.5 mg per 100 g sample, 962.8 mg per 100 g sample and 567.5 mg per 100 g sample, respectively, for Minho, Trás-os-Montes and Beira-Alta). The predominant vitamer was γ-tocopherol (670 mg per 100 g sample for Trás-os-Montes). The antimicrobial and antioxidant activities of C. sativa shell were also determined. Trás-os-Montes extracts displayed the highest antioxidant activity (EC50 = 31.8 ± 1.3 μg mL(-1) for DPPH; 8083.5 ± 164.8 μmol per mg db for FRAP). The total phenolic content (TPC) varied from 241.9 mg to 796.8 mg gallic acid equivalents (GAE) per g db sample, the highest TPC being obtained for Trás-os-Montes. The total flavonoid content (TFC) varied from 31.4 to 43.3 mg of catechin equivalents (CEQ) per g db sample. No antimicrobial activity was observed. The results showed the potentialities of C. sativa shell extracts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3251
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
C29H50O2
T3634
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
C29H50O2
T1539
(+)-α-Tocopherol, Type VI, from vegetable oil, neat (liquid, ≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
C29H50O2
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
296309
1,4-Dioxane, anhydrous, 99.8%
C4H8O2
07-5360
1,4-Dioxane, SAJ first grade
C4H8O2
07-5390
1,4-Dioxane, for HPLC
C4H8O2
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6
B1378
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
C15H24O
258024
α-Tocopherol, ≥95.5%
C29H50O2
563919
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
AlCl3
294713
Aluminum chloride, 99.99% trace metals basis
AlCl3
449598
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
AlCl3
W218405
Butylated hydroxytoluene, ≥99%, FCC, FG
C15H24O
G7384
Gallic acid, 97.5-102.5% (titration)
C7H6O5
451045
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
I2
229695
Iodine, 99.999% trace metals basis
I2
326143
Iodine, ≥99.99% trace metals basis
I2
577782
Sodium carbonate decahydrate, 99.999% trace metals basis
CNa2O3 · 10H2O
563218
Sodium nitrite, 99.999% trace metals basis
NNaO2