EMAIL THIS PAGE TO A FRIEND
Advanced synthesis & catalysis
Asymmetric Synthesis of Substituted Thiolanes through Domino Thia-Michael-Henry Dynamic Covalent Systemic Resolution using Lipase Catalysis.
PMID 26190961
Abstract
Dynamic systems based on consecutive thia-Michael and Henry reactions were generated and transformed using lipase-catalyzed asymmetric transformation. Substituted thiolane structures with three contiguous stereocenters were resolved in the process in high yields and high enantiomeric excesses.
Related Materials