EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
MiR-26b inhibits hepatocellular carcinoma cell proliferation, migration, and invasion by targeting EphA2.
PMID 26191168
Abstract
Deregulated microRNAs (miRNAs) have been shown to play important roles in cancer progression as a result of changes in expression of their target genes. In this study, we investigated the expression of miR-16b in eight hepatocellular carcinoma (HCC) cell lines, revealed the roles of miR-26b on hepatocellular carcinoma (HCC) cell proliferation, migration, and invasion, and confirmed that EphA2 is a direct target of miR-26b. The miR-26b expression was decreased and EphA2 expression was evaluated in HCC cell lines. Luciferase assays revealed that miR-26b inhibited EphA2 expression by targeting the 3'-untranslated region of EphA2 mRNA. Overexpression of miR-26b dramatically inhibited the proliferation, invasion, and migration of HCC cells by targeting EphA2. Moreover, miR-26b down-regulated c-Myc and CyclinD1 expression, which was reversed by overexpressed EphA2. Taken together, our data demonstrated the mechanism of miR-26b contributed to HCC progression and implicated that miR-26b's potential in HCC therapy.