EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Bevacizumab induces A549 cell apoptosis through the mechanism of endoplasmic reticulum stress in vitro.
PMID 26191230
Abstract
To observe the effect of bevacizumab on human A549 cells and explore its mechanism. After different concentrations (0 μM, 1 μM, 5 μM, 25 μM) of bevacizumab treating in A549 cells, CCK8 assay detect the impact of bevacizumab on A549 cell proliferation and flow cytometry determine the effect of bevacizumab on human A549 cells apoptosis. Real-time PCR and Western blotting detect the changing expression of the target gene (CHOP, caspase-4, IRE1, XBP-1) on mRNA and Protein level. Treatment with bevacizumab for 24-hr have induced cell death in a does-dependent manner dramatically (P<0.05). In terms of the mRNA level, expression of XBP-1 has increased obviously in each group (1 μM, 5 μM, 25 μM) (P<0.01); the expression of CHOP (25 μM) and caspase-4 (5 μM) have increased slightly (P<0.05). In terms of the protein level, the expression of CHOP has increased obviously in each group (1 μM, 5 μM, 25 μM) when compared with the control group (0 μM) (P<0.05). As for caspase-4 (5 μM, 25 μM), the expression have increased slightly when compared with the control group (0 μM) (P<0.05). Bevacizumab can induce A549 cell apoptosis through the mechanism of endoplasmic reticulum stress.