PloS one
Maternal Caffeine Consumption during Pregnancy and Risk of Low Birth Weight: A Dose-Response Meta-Analysis of Observational Studies.
PMID 26193706
Abstract
Epidemiologic studies have shown inconsistent conclusions about the effect of caffeine intake during pregnancy on the risk of low birth weight (LBW). We performed a meta-analysis and linear-dose response analysis examining the association between caffeine consumption during pregnancy and risk of LBW. PubMed and EMBASE were searched for relevant articles published up to March 2014. Eight cohort and four case-control studies met all inclusion criteria. Using a random-effects model of the twelve studies, the pooled odds ratio (OR) for the risk of LBW comparing the highest versus lowest level of caffeine intake during pregnancy was 1.38 (95% CI: 1.10, 1.73). Linear dose-response analysis showed that every additional 100 mg of caffeine intake (1 cup of coffee or 2 cups of tea) per day during pregnancy was associated with a 3.0% increase in OR for LBW. There was a moderate level of overall heterogeneity with an I-squared value of 55% (95% CI: 13, 76%), and no evidence of publication bias based on Egger's test (P = 0.20) and the funnel plot. Thus, high caffeine intake during pregnancy is associated with a significant increase in the risk of LBW, and this risk appears to increase linearly as caffeine intake increases.