EMAIL THIS PAGE TO A FRIEND
BMC cancer
Nrf2 is a potential prognostic marker and promotes proliferation and invasion in human hepatocellular carcinoma.
PMID 26194347
Abstract
Nuclear factor E2-related factor 2 (Nrf2 or NFE2L2) is abundantly expressed in cancer cells and relates to proliferation, invasion, and chemoresistance. Our early observations also found that expression of Nrf2 was up-regulated in kinds of cancer including human hepatocellular carcinoma (HCC) cells. But there are limited reports about the expression, significance, function of Nrf2 in HCC. First, Nrf2 expression was analyzed in HCC cell lines and tumor samples. Then, the relationship of Nrf2 with clinicopathological factors and survival were analyzed. Further, the effect of Nrf2 on cell proliferation, apoptosis, and metastasis was examined in vitro by modulating expression of Nrf2 through specific shRNA or expression plasmid. Last, the potential mechanisms were also investigated. Nrf2 was up-regulated in HCC, and expression of Nrf2 was correlated with tumor differentiation, metastasis, and tumor size. Univariate and multivariate analyses indicated that high Nrf2 expression might be a poor prognostic factor. Further studies demonstrated that inhibition of Nrf2 expression inhibited proliferation by inducing apoptosis and repressed invasion, and up-regulation of Nrf2 expression resulted in opposite phenotypes. Moreover, there are positive correlation between Nrf2 expression and that of Bcl-xL and MMP-9. Nrf2 is a potential prognostic marker and promotes proliferation and invasion in human hepatocellular carcinoma partly through regulating expression of Bcl-xL and MMP-9.