EMAIL THIS PAGE TO A FRIEND
Diagnostic pathology
The microRNA-325 inhibits hepatocellular carcinoma progression by targeting high mobility group box 1.
PMID 26194496
Abstract
MicroRNAs (miRNAs) can serve as tumor suppressors and might provide an efficient strategy for annihilating tumor cells. Nevertheless, the potential role of miR-325 in hepatocellular carcinoma (HCC) is still unknown. Using RT-PCR, immunoblots invasion assays and bioinformatics strategies, we investigated the potential role of miR-325 in HCC. We showed that miR-325 was decreased and HMGB1 was increased in 99 patients with hepatocellular carcinoma. MiR-325 inhibition promoted cell invasion and proliferation, while miR-325 upregulation inhibited cell invasion and proliferation by using transwell and CCK8 assays. We further showed that HMGB1 might be a direct target of miR-325 and is negatively regulated by miR-325. Down-regulation of miR-325 predicts poor prognosis for HCC patients. These findings implied that miR-325 regulates cell invasion and proliferation via targeting HMGB1 and may be a potential prognostic marker for HCC. The virtual slide(s) for this article can be found here: http://www.diagnosticpathology.diagnomx.eu/vs/4655707031717989.