EMAIL THIS PAGE TO A FRIEND
Experimental Brain Research. Experimentelle Hirnforschung. Experimentation Cerebrale
Chronic deep brain stimulation of the medial forebrain bundle reverses depressive-like behavior in a hemiparkinsonian rodent model.
PMID 26195164