EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Restoration of CD4+ Responses to Copathogens in HIV-Infected Individuals on Antiretroviral Therapy Is Dependent on T Cell Memory Phenotype.
PMID 26195814
Abstract
Antiretroviral therapy (ART) induces rapid suppression of viral replication and a progressive replenishment of CD4(+) T cells in HIV-infected individuals. However, the effect of ART on restoring pre-existing memory CD4(+) T cells specific for common copathogens is still unclear. To better understand the dynamics of Ag-specific CD4(+) T cells during ART, we assessed the frequency, functional capacity, and memory profile of CD4(+) T cells specific for Mycobacterium tuberculosis and CMV in 15 HIV-infected individuals before and 1 y after ART initiation. After ART initiation, the frequency of M. tuberculosis-specific CD4(+) T cells showed little change, whereas CMV-specific CD4(+) T cells were significantly lower (p = 0.003). There was no difference in the polyfunctional or memory profile of Ag-specific CD4(+) T cells before and after ART. The replenishment of Ag-specific CD4(+) T cells correlated with the memory differentiation profile of these cells prior to ART. Pathogen-specific CD4(+) T cells exhibiting a late differentiated profile (CD45RO(+)CD27(-)) had a lower capacity to replenish (p = 0.019; r = -0.5) compared with cells with an early differentiated profile (CD45RO(+)CD27(+); p = 0.04; r = 0.45). In conclusion, restoration of copathogen-specific memory CD4(+) T cells during treated HIV infection is related to their memory phenotype, in which early differentiated cells (such as most M. tuberculosis-specific cells) have a higher replenishment capacity compared with late differentiated cells (such as most CMV-specific cells). These data identify an important, hitherto unrecognized, factor that may limit restoration of copathogen immunity in HIV-infected individuals on ART.