EMAIL THIS PAGE TO A FRIEND
Environmental Research
Lead concentrations in whole blood, serum, saliva and house dust in samples collected at two time points (12 months apart) in Santo Amaro, BA, Brazil.
PMID 26196778