EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Accurate measurement of the essential micronutrients methionine, homocysteine, vitamins B6, B12, B9 and their metabolites in plasma, brain and maternal milk of mice using LC/MS ion trap analysis.
PMID 26197436
Abstract
Methionine, homocysteine, vitamins B6, B12, B9, and their metabolites are crucial co-factors and substrates for many basic biological pathways including one-carbon metabolism, and they are particularly important for brain function and development and epigenetic mechanisms. These are essential nutrients that cannot be synthesized endogenously and thus need to be taken in via diet. A novel method was developed that enables simultaneous assessment of the exact concentrations of these essential micronutrients in various matrices, including maternal milk, plasma, and brain of neonatal mice. The protocol for analysis of these components in the various matrices consists of a cleanup step (i.e. lipid extraction followed by protein precipitation) combined with a liquid chromatography mass spectrometry (LC/MS) ion trap method with high sensitivity and selectivity (SRM mode). This novel method enables the measurement of these essential nutrients with good recoveries (69-117%), and high intra-day (<10%) and high intra-day precision (defined as <15% for compounds with an isotopologue and <20% for compounds without an isotopologue as internal standard) in plasma, maternal milk, and brain of mice at low and high levels. In addition, lower limits of quantitation (LOQ) were determined for the various matrices in the range for methionine (700-2000nmol/L), homocysteine (280-460-nmol/L), vitamins B6 (5-230nmol/L), B12 (7-11nmol/L), B9 (20-30nmol/L). Degradation of vitamins and oxidation of homocysteine is limited to a minimum, and only small sample volumes (30μL plasma, 20mg brain and maternal milk) are needed for simultaneous measurement. This method can help to understand how these nutrients are transferred from mother to offspring via maternal milk, as well as how these nutrients are absorbed by the offspring and eventually taken up in various tissues amongst the brain in preclinical and clinical research settings. Therefore the method can help to explore critical periods in lactating mothers and developing offspring.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8255
1,4-Dithioerythritol, ≥99.0%
C4H10O2S2
D9680
1,4-Dithioerythritol, BioXtra, ≥99.0%
C4H10O2S2
D8161
1,4-Dithioerythritol, BioReagent, for molecular biology, ≥99.0%
C4H10O2S2
43794
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
C4H10O2S2
443751
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%
C7H5F7O2
C3607
Cyanocobalamin, meets USP testing specifications
C63H88CoN14O14P
F7876
Folic acid, ≥97%
C19H19N7O6
F8758
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
C19H19N7O6
F8798
Folic acid, meets USP testing specifications
C19H19N7O6
77249
Heptafluorobutyric acid, ≥99.0% (GC)
C4HF7O2
164194
Heptafluorobutyric acid, 98%
C4HF7O2
M9625
L-Methionine, reagent grade, ≥98% (HPLC)
C5H11NO2S
M5308
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
C5H11NO2S
M8439
L-Methionine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C5H11NO2S
64319
L-Methionine, BioUltra, ≥99.5% (NT)
C5H11NO2S
P5669
Pyridoxine, ≥98%
C8H11NO3
V2876
Vitamin B12, ≥98%
C63H88CoN14O14P
V6629
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
C63H88CoN14O14P