EMAIL THIS PAGE TO A FRIEND
Natural product communications
A New Aromatic Compound from the Stem Bark of Terminalia catappa.
PMID 26197537
Abstract
A new aromatic compound 3,4,5-trimethoxyphenyl-1-O-(4-sulfo)-β-D-glucopyranoside (1), in addition to two triterpenoid saponins (chebuloside II, arjunoglucoside II), two triterpenes (arjunolic acid and 3-betulinic acid) and sitosterol-3-O-β-D-glucopyranoside have been isolated from the barks of Terminalia catappa. Their structures have been established on the basis of spectroscopic techniques (1D/2D NMR) and MS. Their cytotoxicity and antiinflammatory activity, together with the antioxidant capacity of compound 1 were also evaluated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149691
Thiazolidine, 95%
C3H7NS