EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Glass and Plastics Platforms for Foldable Electronics and Displays.
PMID 26197761
Abstract
Reversibly, repeatedly foldable electronics and displays are enabled by employing engineered glass or plastics substrates, where folding deformation is localized in thinned parts only. This design concept can further be extended to dual folding, leading to size reduction down to 1/4. Notably, the foldable electronics and displays can be implemented with no need to introduce any novel materials.