EMAIL THIS PAGE TO A FRIEND
Daru : journal of Faculty of Pharmacy, Tehran University of Medical Sciences
Phthalimide analogs as probable 15-lipoxygenase-1 inhibitors: synthesis, biological evaluation and docking studies.
PMID 26197788
Abstract
Recent studies have been explained the role of lipoxygenases (LOX) in the origin of cancer. Among the lipoxygenases, the 5-LOX, 12-LOX and 15-LOX are more important in the cause of neoplastic disorders. In the present investigation, a new series of anticancer agents with 1,3,4-thiadiazole and phthalimide substructures were synthesized and their in vitro cytotoxicity was evaluated by MTT assay. Moreover, enzyme inhibitory potency was also assessed by enzymatic protocol towards 15-LOX-1. Molecular docking was performed to explore in silico binding mode of the target compounds. Tested compounds showed a better cytotoxic activity against HT29 cell line (colorectal cancer) in comparison with other cell lines (PC3: prostate carcinoma; SKNMC: neuroblastoma). Unfortunately, all of the tested derivatives rendered lower inhibitory potency than quercetin towards 15-LOX-1. Four hydrogen bonds were detected in docking studies for compound 4d as the most potent derivative in enzymatic assay. The biological results of reported compounds in this research were not so satisfactory. But, further structural modifications are necessary to improve the bioactivity of these derivatives.