EMAIL THIS PAGE TO A FRIEND
Nutrients
A Dietary Pattern Derived by Reduced Rank Regression is Associated with Type 2 Diabetes in An Urban Ghanaian Population.
PMID 26198248
Abstract
Reduced rank regression (RRR) is an innovative technique to establish dietary patterns related to biochemical risk factors for type 2 diabetes, but has not been applied in sub-Saharan Africa. In a hospital-based case-control study for type 2 diabetes in Kumasi (diabetes cases, 538; controls, 668) dietary intake was assessed by a specific food frequency questionnaire. After random split of our study population, we derived a dietary pattern in the training set using RRR with adiponectin, HDL-cholesterol and triglycerides as responses and 35 food items as predictors. This pattern score was applied to the validation set, and its association with type 2 diabetes was examined by logistic regression. The dietary pattern was characterized by a high consumption of plantain, cassava, and garden egg, and a low intake of rice, juice, vegetable oil, eggs, chocolate drink, sweets, and red meat; the score correlated positively with serum triglycerides and negatively with adiponectin. The multivariate-adjusted odds ratio of type 2 diabetes for the highest quintile compared to the lowest was 4.43 (95% confidence interval: 1.87-10.50, p for trend < 0.001). The identified dietary pattern increases the odds of type 2 diabetes in urban Ghanaians, which is mainly attributed to increased serum triglycerides.