EMAIL THIS PAGE TO A FRIEND
Andrology
Sperm Lysozyme-Like Protein 1 (SLLP1), an intra-acrosomal oolemmal-binding sperm protein, reveals filamentous organization in protein crystal form.
PMID 26198801
Abstract
Sperm lysozyme-like protein 1 (SLLP1) is one of the lysozyme-like proteins predominantly expressed in mammalian testes that lacks bacteriolytic activity, localizes in the sperm acrosome, and exhibits high affinity for an oolemmal receptor, SAS1B. The crystal structure of mouse SLLP1 (mSLLP1) was determined at 2.15xa0Å resolution. mSLLP1 monomer adopts a structural fold similar to that of chicken/mouse lysozymes retaining all four canonical disulfide bonds. mSLLP1 is distinct from c-lysozyme by substituting two essential catalytic residues (E35T/D52N), exhibiting different surface charge distribution, and by forming helical filaments approximately 75xa0Å in diameter with a 25xa0Å central pore comprised of six monomers per helix turn repeating every 33xa0Å. Cross-species alignment of all reported SLLP1 sequences revealed a set of invariant surface regions comprising a characteristic fingerprint uniquely identifying SLLP1 from other c-lysozyme family members. The fingerprint surface regions reside around the lips of the putative glycan-binding groove including three polar residues (Y33/E46/H113). A flexible salt bridge (E46-R61) was observed covering the glycan-binding groove. The conservation of these regions may be linked to their involvement in oolemmal protein binding. Interaction between SLLP1 monomer and its oolemmal receptor SAS1B was modeled using protein-protein docking algorithms, utilizing the SLLP1 fingerprint regions along with the SAS1B conserved surface regions. This computational model revealed complementarity between the conserved SLLP1/SAS1B interacting surfaces supporting the experimentally observed SLLP1/SAS1B interaction involved in fertilization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
792527
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
C3H4N2
I5513
Imidazole, for molecular biology, ≥99% (titration)
C3H4N2
I0250
Imidazole, ≥99% (titration), crystalline
C3H4N2
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2
56750
Imidazole, puriss. p.a., ≥99.5% (GC)
C3H4N2
I202
Imidazole, ReagentPlus®, 99%
C3H4N2
56748
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
C3H4N2
56749
Imidazole, BioUltra, ≥99.5% (GC)
C3H4N2
68268
Imidazole buffer Solution, BioUltra, 1 M in H2O
C3H4N2
451045
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
I2
229695
Iodine, 99.999% trace metals basis
I2
326143
Iodine, ≥99.99% trace metals basis
I2
N0128
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
C6H7NNa2O6
N0253
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
C6H6NNa3O6
U0631
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
CH4N2O
U5378
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
CH4N2O
U4883
Urea, 8 M (after reconstitution with 16 mL high purity water)
CH4N2O
U6504
Urea, for electrophoresis
CH4N2O
U4884
Urea, meets USP testing specifications
CH4N2O
U4128
Urea, BioReagent, suitable for cell culture
CH4N2O
U5128
Urea, ACS reagent, 99.0-100.5%
CH4N2O
U1250
Urea, ReagentPlus®, ≥99.5%, pellets
CH4N2O
51456
Urea, BioUltra, for molecular biology, ≥99.5% (T)
CH4N2O
33247
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
CH4N2O
15604
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
CH4N2O
U7129
Urea solution, 40 % (w/v) in H2O
CH4N2O
51457
Urea solution, BioUltra, ~8 M in H2O
CH4N2O
299367
Urea-12C, 99.9 atom % 12C
12CH4N2O
608734
Urea-14N2, 99.99 atom % 14N, 15N-depleted
CH414N2O