EMAIL THIS PAGE TO A FRIEND
Laboratory medicine
Associations Between Inflammatory Markers, Hemostatic Markers, and Microvascular Complications in 182 Chinese Patients With Type 2 Diabetes Mellitus.
PMID 26199261
Abstract
To examine the associations between inflammatory markers, coagulation and fibrinolysis parameters, and microvascular complications in 182 Chinese patients with type 2 diabetes mellitus (T2DM) who sought treatment at a large hospital in Zhejiang province, China. We investigated the relationships of blood inflammatory markers with hemostatic markers in 87 patients with T2DM who did not have complications and 95 patients with T2DM who had microvascular complications. C-reactive protein (CRP) and interleukin-6 (IL-6) were significantly correlated with fibrinogen, thrombin-antithrombin III complex (TAT III), plasminogen activator inhibitor-1 (PAI-1), von Willebrand factor (vWF), and coagulation factors (F) VII in patients with T2DM who had microvascular complications (P <.05). Based on logistic regression analysis, the highest-tertile groups of fibrinogen, FVII, and FVIII, corresponded to a greater risk of high CRP, whereas risk of high IL-6 was significantly greater in the groups with highest-tertile values for fibrinogen, FVII, TAT III, PAI-1, and activated protein C (APC). Elevated levels of CRP and IL-6 might be associated with increased coagulability and a tendency towards thrombus formation in patients with T2DM who have microvascular complications.