PloS one
Predictive and Prognostic Value of Ribonucleotide Reductase Regulatory Subunit M1 and Excision Repair Cross-Complementation Group 1 in Advanced Urothelial Carcinoma (UC) Treated with First-Line Gemcitabine Plus Platinum Combination Chemotherapy.
PMID 26200905
Abstract
Preclinical and clinical studies have suggested that expression of ribonucleotide reductase regulatory subunit M1 (RRM1) and excision repair cross-complementation group 1 (ERCC1) is associated with resistance to gemcitabine and cisplatin, respectively. We evaluated the significance of RRM1 and ERCC1 expression to predict tumor response to gemcitabine plus platinum chemotherapy (GP) and survival in advanced UC. We retrospectively collected tumor samples and reviewed clinical data of 53 patients with unresectable or metastatic UC, who were treated with first-line GP. RRM1 and ERCC1 expression were measured by immunohistochemistry. Among 53 patients, 12 (22.6%) and 26 (49.1%) patients had tumors that demonstrated a high expression for RRM1 and ERCC1, respectively. Twenty-nine (70.7%) of 41 patients with low RRM1 expression achieved a clinical response (complete + partial responses), but only 3 (25.0%) of 12 patients with high RRM1 expression achieved a clinical response after GP (P=0.007). Nineteen (70.4%) of 27 patients with low ERCC1 expression achieved a clinical response, while 13 (50.0%) of 26 patients with high ERCC1 expression achieved a clinical response (P=0.130). High RRM1 expression was associated with shorter progression free survival and overall survival (PFS P=0.006, OS P=0.006). Multivariate analysis confirmed that patients with high RRM1 expression had a significantly greater risk of progression and death than those with low RRM1 expression. ERCC1 status was not a significant predictor for PFS and OS. RRM1 expression was predictive and prognostic of clinical outcome in advanced UC treated with gemcitabine plus platinum combination chemotherapy.