EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Sensitizing potential of cyclobutenediones.
PMID 2620514
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
310778
3,4-Diethoxy-3-cyclobutene-1,2-dione, 98%
C8H10O4