EMAIL THIS PAGE TO A FRIEND
Psychological science
Creativity and Memory: Effects of an Episodic-Specificity Induction on Divergent Thinking.
PMID 26205963
Abstract
People produce more episodic details when imagining future events and solving means-end problems after receiving an episodic-specificity induction-brief training in recollecting details of a recent event-than after receiving a control induction not focused on episodic retrieval. Here we show for the first time that an episodic-specificity induction also enhances divergent creative thinking. In Experiment 1, participants exhibited a selective boost on a divergent-thinking task (generating unusual uses of common objects) after a specificity induction compared with a control induction; by contrast, performance following the two inductions was similar on an object association task thought to involve little divergent thinking. In Experiment 2, we replicated the specificity-induction effect on divergent thinking using a different control induction, and also found that participants performed similarly on a convergent-thinking task following the two inductions. These experiments provide novel evidence that episodic memory is involved in divergent creative thinking.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674397
11-Amino-1-undecanethiol hydrochloride, 99%
C11H25NS · HCl