EMAIL THIS PAGE TO A FRIEND
AIDS (London, England)
Interacting partners of macrophage-secreted cathepsin B contribute to HIV-induced neuronal apoptosis.
PMID 26208400
Abstract
HIV-1 infection of macrophages increases cathepsin B secretion and induces neuronal apoptosis, but the molecular mechanism remains unclear. We identified macrophage-secreted cathepsin B protein interactions extracellularly and their contribution to neuronal death in vitro. Cathepsin B was immunoprecipitated from monocyte-derived macrophage supernatants after 12 days postinfection. The cathepsin B interactome was identified by label-free tandem mass spectrometry and compared with uninfected supernatants. Proteins identified were validated by western blot. Neurons were exposed to macrophage-conditioned media in presence or absence of antibodies against cathepsin B and interacting proteins. Apoptosis was measured using TUNEL labeling. Immunohistochemistry of postmortem brain tissue samples from healthy, HIV-infected and Alzheimer's disease patients was performed to observe the ex-vivo expression of the proteins identified. Nine proteins co-immunoprecipitated differentially with cathepsin B between uninfected and HIV-infected macrophages. Serum amyloid P component (SAPC)-cathepsin B interaction increased in HIV-infected macrophage supernatants, while matrix metalloprotease 9 (MMP-9)-cathepsin B interaction decreased. Pre-treatment of HIV-infected macrophage-conditioned media with antibodies against cathepsin B and SAPC decreased neuronal apoptosis. The addition of MMP-9 antibodies was not neuro-protective SAPC was overexpressed in postmortem brain tissue from HIV-positive neurocognitive impaired patients compared with HIV positive with normal cognition and healthy controls, although MMP-9 expression was similar in all tissues. Inhibiting SAPC-cathepsin B interaction protects against HIV-induced neuronal death and may help to find alternative treatments for HIV-associated neurocognitive disorders.