EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Evaluation of the matrix effect of different sample matrices for 33 pharmaceuticals by post-column infusion.
PMID 26210586
Abstract
Matrix effects that occur during quantitative measurement by liquid chromatography mass spectrometry specifically when using electrospray ionization are a widely recognized phenomenon. Sample matrix compounds affect the ionization process of the target analytes, lead to a low signal response, and flawed analytical results. How these matrix compounds directly influence the ionization process has not yet been completely understood. In the present study, we determined the matrix effect for 33 pharmaceutical substances in sample extracts of urine, plasma and wastewater. Most of the investigated substances were subject to a signal suppression effect. Only for a small subset of the compounds we detected a signal enhancement effect. We investigated the matrix effect profiles in detail to disentangle the influence of different matrices and to correlate the impact of specific components and groups of the analyzed extract in suppressing or enhancing effects in the profile. Most signal suppression effects were detected in the first half of the chromatographic run-time for the matrix extracts of urine and wastewater. The observed effects are caused by high mass flow of salts and other diverse matrix components that were contained in high concentrations in those biological matrices. We also found signal suppression in the matrix effect profile of plasma samples over a wide time range during the chromatographic separation that were associated with a high content of triglycerides of diverse carbohydrate chain lengths. Here, we provide a broader picture of how 33 substances were influenced during analysis. Our results imply that a high number of the investigated substances had comparable effects of matrix compounds, despite differences in their chemical structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D-091
10,11-Dihydro-10-Hydroxycarbamazepine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
C15H14N2O2
A-072
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
C14H22N2O3
C8981
Carbamazepine, meets USP testing specifications
C15H12N2O
C-053
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C15H12N2O
C6305
Clozapine
C18H19ClN4
C171
Clozapine, solid
C18H19ClN4
C-059
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C18H19ClN4
F-031
Fluconazole solution, 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C13H12F2N6O
G154
Gabapentin, solid
C9H17NO2
G-007
Gabapentin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C9H17NO2
G007
Guvacine hydrochloride, ≥97% (NMR), solid
C6H9NO2 · HCl
H100
Haloperidol
C21H23ClFNO2
H-030
Haloperidol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C21H23ClFNO2
L3791
Lamotrigine, ≥98%, powder
C9H7Cl2N5
L-019
Lamotrigine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C9H7Cl2N5
L8668
Levetiracetam, ≥98% (HPLC)
C8H14N2O2
L-020
Levetiracetam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C8H14N2O2
M0443
Mirtazapine, ≥98% (HPLC)
C17H19N3
M-128
Mirtazapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C17H19N3
V-007
O-Desmethylvenlafaxine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C16H25NO2
O1141
Olanzapine, ≥98% (HPLC)
C17H20N4S
O-024
Olanzapine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
C17H20N4S
O3764
Oxcarbazepine, ≥98% (HPLC), solid
C15H12N2O2
PZ0010
Pregabalin, ≥97% (NMR)
C8H17NO2
P-066
Pregabalin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C8H17NO2
P-075
Primidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C12H14N2O2
R-006
Risperidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C23H27FN4O2
T-039
Topiramate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C12H21NO8S
T-904
Trimipramine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C20H26N2