EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Controllable Chemoselectivity in Visible-Light Photoredox Catalysis: Four Diverse Aerobic Radical Cascade Reactions.
PMID 26212794
Abstract
Reported is the controllable selectivity syntheses of four distinct products from the same starting materials by visible-light photoredox catalysis. By employing a dicyanopyrazine-derived chromophore (DPZ) as photoredox catalyst, an aerobic radical mechanism has been developed, and allows the reactions of N-tetrahydroisoquinolines (THIQs) with N-itaconimides to through four different pathways, including addition-cyclization, addition-elimination, addition-coupling, and addition-protonation, with satisfactory chemoselectivity. The current strategy provide straightforward access to four different but valuable N-heterocyclic adducts in moderate to excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
908444
DPZ, ≥95%
C16H10N4O2S2